missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylcatechol, 98%

Product Code. 11314739
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11314739 250 mg 250mg
11439922 1 g 1g
11449922 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314739 Supplier Thermo Scientific Alfa Aesar Supplier No. A12048.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1124-39-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-39-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00015847
InChI Key HFLGBNBLMBSXEM-UHFFFAOYSA-N
Synonym 4-ethylcatechol, 4-ethylpyrocatechol, 4-ethyl-1,2-benzenediol, unii-574jv8byr2, 1,2-benzenediol, 4-ethyl, benzenediol, 4-ethyl, 3,4-dihydroxyethylbenzene, ethylcatechol, 4-ethyl catechol, 4-ethylc atechol
PubChem CID 70761
IUPAC Name 4-ethylbenzene-1,2-diol
SMILES CCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 37°C to 40°C
Boiling Point 105°C to 107°C (0.5 mmHg)
Quantity 250 mg
Beilstein 2042062
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Ethylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.