missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzylamine, 98%, Thermo Scientific Chemicals

Product Code. 10310254 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10310254 5 g Glass bottle 5g
10551534 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310254 Supplier Thermo Scientific Supplier No. 391140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7441-43-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD01529871
InChI Key DGAGEFUEKIORSQ-UHFFFAOYSA-N
Synonym 4-ethylbenzylamine, 4-ethylphenyl methanamine, 1-4-ethylphenyl methanamine, 4-ethylphenyl methylamine, 4-ethyl benzylamine, 4-ethyl-benzylamine, 4-ethyl-benzyl amine, benzenemethanamine, 4-ethyl
PubChem CID 3870219
IUPAC Name (4-ethylphenyl)methanamine
SMILES CCC1=CC=C(C=C1)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8830g/mL
Boiling Point 208°C (700.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2C6H4CH2NH2
Refractive Index 1.5280 to 1.53
Quantity 5 g
Beilstein 12, IV, 2641
Specific Gravity 0.883
Solubility Information Solubility in water: soluble
Formula Weight 135.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.