missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl chloride, 97%, stab. with calcium carbonate, Thermo Scientific™

Product Code. p-7030323
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15477735

Brand: Thermo Scientific Alfa Aesar H27900.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1467-05-6
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00039357
InChI Key DUBCVXSYZVTCOC-UHFFFAOYSA-N
Synonym 4-ethylbenzyl chloride, 1-chloromethyl-4-ethylbenzene, p-ethylbenzylchloride, p-ethylbenzyl chloride, benzene, 1-chloromethyl-4-ethyl, ethyxlbenzylchloride, 4-ethylbenzylchloride, 1-chloromethyl-4-ethyl-benzene, 4-chloromethyl-1-ethylbenzene, 4-ethylbenzyl chloride contains ca 2-form
PubChem CID 73843
IUPAC Name 1-(chloromethyl)-4-ethylbenzene
SMILES CCC1=CC=C(CCl)C=C1

Specifications

Density 1.04
Boiling Point 111°C (26 mmHg)
Quantity 5 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 154.64
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzyl chloride, stabilized with calcium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.