missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl chloride, 97%, stab. with calcium carbonate, Thermo Scientific™

Product Code. 15487735
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15487735 25 g 25g
15477735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487735 Supplier Thermo Scientific Alfa Aesar Supplier No. H27900.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1467-05-6
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00039357
InChI Key DUBCVXSYZVTCOC-UHFFFAOYSA-N
Synonym 4-ethylbenzyl chloride, 1-chloromethyl-4-ethylbenzene, p-ethylbenzylchloride, p-ethylbenzyl chloride, benzene, 1-chloromethyl-4-ethyl, ethyxlbenzylchloride, 4-ethylbenzylchloride, 1-chloromethyl-4-ethyl-benzene, 4-chloromethyl-1-ethylbenzene, 4-ethylbenzyl chloride contains ca 2-form
PubChem CID 73843
IUPAC Name 1-(chloromethyl)-4-ethylbenzene
SMILES CCC1=CC=C(CCl)C=C1

Specifications

Density 1.04
Boiling Point 111°C (26 mmHg)
Quantity 25 g
UN Number UN3265
Solubility Information Insoluble in water.
Formula Weight 154.64
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzyl chloride, stabilized with calcium carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.