missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzyl alcohol, 99%

Product Code. 15433947
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15433947

Brand: Thermo Scientific Alfa Aesar H30459.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-59-2 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-59-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004666
InChI Key YSLBFFIVJGJBSA-UHFFFAOYSA-N
PubChem CID 13034
IUPAC Name (4-ethylphenyl)methanol
SMILES CCC1=CC=C(C=C1)CO

Specifications

Density 1.028
Boiling Point 115°C to 117°C (9 mmHg)
Refractive Index 1.5275
Quantity 5 g
Synonym 4-ethylphenyl methanol, 4-ethylbenzyl alcohol, 4-ethylbenzylalcohol, benzenemethanol, 4-ethyl, benzyl alcohol, p-ethyl, p-ethylbenzyl alcohol, 4-ethyl-benzyl alcohol, benzenemethanol,4-ethyl, acmc-209p6g, 4-ethylphenyl methanol #
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 4-Ethylbenzyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.