missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzoyl chloride, 98+%

Product Code. 11419213
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419213 10 g 10g
11324739 5 g 5g
11429213 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419213 Supplier Thermo Scientific Alfa Aesar Supplier No. A11698.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16331-45-6 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16331-45-6
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000697
InChI Key AVTLLLZVYYPGFX-UHFFFAOYSA-N
PubChem CID 85368
IUPAC Name 4-ethylbenzoyl chloride
SMILES CCC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.146
Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Odor Acidic
Refractive Index 1.5479
Quantity 10 g
UN Number UN3265
Beilstein 2206041
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 4-ethyl, p-ethylbenzoyl chloride, 4-ethylbenzoylchloride, 4-ethylbenzene-1-carbonyl chloride, 4-ethyl-benzoyl chloride, acmc-20amiy, pubchem9725, 4-ethyl benzoyl choride, 4-ethyl benzoyl chloride, benzoyl chloride,4-ethyl
Formula Weight 168.62
Percent Purity ≥98%
Chemical Name or Material 4-Ethylbenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.