missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11382097
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11382097 10 g 10g
11392097 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382097 Supplier Thermo Scientific Alfa Aesar Supplier No. L07921.09
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25309-65-3 | Molecular Formula: C9H9N | Molecular Weight: 131.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25309-65-3
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00016383
InChI Key SXFFMFAQNAFSLF-UHFFFAOYSA-N
Synonym benzonitrile, 4-ethyl, p-ethylbenzonitrile, 1-cyano-4-ethylbenzene, 4-ethylbenzenecarbonitrile, 4-ethyl-benzonitrile, acmc-20amj0, 4-ethylbenzonitrile
PubChem CID 91393
IUPAC Name 4-ethylbenzonitrile
SMILES CCC1=CC=C(C=C1)C#N

Specifications

Density 0.956
Boiling Point 237°C
Flash Point 97°C (206°F)
Refractive Index 1.526
Quantity 10 g
Beilstein 2040912
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 4-Ethylbenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.