missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzoic acid, 97%

Product Code. 10042420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042420 25 g Glass bottle 25g
10040540 25 g Glass bottle Each
10563741 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10042420 Supplier Thermo Scientific Acros Supplier No. 184490250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-64-7 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-64-7
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002570
InChI Key ZQVKTHRQIXSMGY-UHFFFAOYSA-N
Synonym benzoic acid, 4-ethyl, p-ethylbenzoic acid, 4-ethyl benzoic acid, 4-ethyl-benzoic acid, unii-9tkm8p0g6r, 9tkm8p0g6r, p-ethylbenzoate, 4-ethyl benzoate, 4-ethylbenzoicacid, p-ethylbenzoic acid;
PubChem CID 12086
IUPAC Name 4-ethylbenzoic acid
SMILES CCC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C2H5C6H4CO2H
Quantity 25 g
Beilstein 09, 529
Solubility Information Solubility in water: slightly soluble
Formula Weight 150.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Ethylbenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.