missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzenesulfonyl chloride, 97%

Product Code. 11490548
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490548 5 g 5g
11400558 25 g 25g
11410558 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490548 Supplier Thermo Scientific Alfa Aesar Supplier No. B24682.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16712-69-9 | C8H9ClO2S | 204.668 g/mol

4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Reacts with water. Soluble in ethanol, acetone and toluene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents, bases, amines and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16712-69-9
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00041480
InChI Key LACFLXDRFOQEFZ-UHFFFAOYSA-N
Synonym 4-ethylbenzene-1-sulfonyl chloride, p-ethylbenzenesulfonyl chloride, 4-ethylbenzenesulphonyl chloride, 4-ethyl-benzenesulfonyl chloride, ethylbenzenesulfonylchloride, 4-ethylbenzenesulfonylchloride, chloro 4-ethylphenyl sulfone, pubchem5133, acmc-209dvn, 4-ethylphenylsulfonyl chloride
PubChem CID 570959
IUPAC Name 4-ethylbenzenesulfonyl chloride
SMILES CCC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 8°C to 12°C
Density 1.268
Boiling Point 134°C to 136°C (7 mmHg)
Refractive Index 1.545
Quantity 5 g
UN Number UN3265
Beilstein 2090130
Sensitivity Moisture sensitive
Solubility Information Reacts with water. Soluble in ethanol,acetone and toluene.
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.