missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10552272
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10552272 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10552272 Supplier Thermo Scientific Maybridge Supplier No. BTB10241DA
 Restricted Item
Estimated Shipment: 01-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16712-69-9
CAS Min % 96.5
CAS Max % 100.0
Density 1.268g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H9ClO2S
Linear Formula CH3CH2C6H4SO2Cl
MDL Number MFCD00041480
Quantity 1 g
Synonym 4-ethylbenzene-1-sulfonyl chloride, p-ethylbenzenesulfonyl chloride, 4-ethylbenzenesulphonyl chloride, 4-ethyl-benzenesulfonyl chloride, ethylbenzenesulfonylchloride, 4-ethylbenzenesulfonylchloride, chloro 4-ethylphenyl sulfone, pubchem5133, acmc-209dvn, 4-ethylphenylsulfonyl chloride
InChI Key LACFLXDRFOQEFZ-UHFFFAOYSA-N
SMILES CCC1=CC=C(C=C1)S(=O)(=O)Cl
IUPAC Name 4-ethylbenzenesulfonyl chloride
Molecular Weight (g/mol) 204.668
PubChem CID 570959
Formula Weight 204.68
Percent Purity 97%
Chemical Name or Material 4-ethylbenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.