missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzaldehyde, 98%

Product Code. 10356992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356992 25 g Glass bottle 25g
10235162 5 g Glass bottle 5g
10432995 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356992 Supplier Thermo Scientific Acros Supplier No. 378950250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4748-78-1 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4748-78-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006956
InChI Key QNGNSVIICDLXHT-UHFFFAOYSA-N
PubChem CID 20861
IUPAC Name 4-ethylbenzaldehyde
SMILES CCC1=CC=C(C=C1)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 221.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2C6H4CHO
Refractive Index 1.5370 to 1.5410
Quantity 25 g
Specific Gravity 0.97
Synonym p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
Formula Weight 134.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.