missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-4-methylmorpholinium Bromide, TRC

Product Code. 30374704
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374704 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374704 Supplier Toronto Research Chemicals Supplier No. TRCE937540500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Ethyl-4-methylmorpholinium Bromide
CAS 65756-41-4
Molecular Formula C7H16BrNO
Quantity 500 mg
InChI Formula InChI=1S/C7H16NO.BrH/c1-3-8(2)4-6-9-7-5-8;/h3-7H2,1-2H3;1H/q+1;/p-1
SMILES [Br-].CC[N+]1(C)CCOCC1
IUPAC Name 4-ethyl-4-methylmorpholin-4-ium;bromide
Molecular Weight (g/mol) 210.11
Formula Weight 209.0415
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.