missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-2,3-dioxo-1-piperazinecarbonyl Chloride, TRC

Product Code. 30388699
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388699 1 g
30370728 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388699 Supplier Toronto Research Chemicals Supplier No. TRCE9173751G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59703-00-3
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
Synonym 1-Piperazinecarbonyl chloride, 4-ethyl-2,3-dioxo-, 4-Ethyl-2,3-dioxo-1-piperazinecarbonyl chloride
SMILES CCN1CCN(C(=O)Cl)C(=O)C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H9ClN2O3/c1-2-9-3-4-10(7(8)13)6(12)5(9)11/h2-4H2,1H3
Formula Weight 204.0302
Chemical Name or Material 4-Ethyl-2,3-dioxo-1-piperazinecarbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.