missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-1,3-dihydro-2H-indol-2-one, TRC

Product Code. 30384995
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384995 10 mg
30374690 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384995 Supplier Toronto Research Chemicals Supplier No. TRCE91732010MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 954117-24-9
Molecular Formula C10 H11 N O
Molecular Weight (g/mol) 161.2
Synonym Ropinirole Impurity C
SMILES CCC1=C2CC(=O)NC2=CC=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H11NO/c1-2-7-4-3-5-9-8(7)6-10(12)11-9/h3-5H,2,6H2,1H3,(H,11,12)
Formula Weight 161.0841
Chemical Name or Material 4-Ethyl-1,3-dihydro-2H-indol-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.