missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenylacetic acid, 98%, Thermo Scientific™

Product Code. 10296150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296150 10 g Glass bottle 10g
10764451 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296150 Supplier Thermo Scientific Acros Supplier No. 156040100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4919-33-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00004346
InChI Key ZVVWZNFSMIFGEP-UHFFFAOYSA-N
Synonym 4-ethoxyphenylacetic acid, 2-4-ethoxyphenyl acetic acid, 4-ethoxyphenyl acetic acid, benzeneacetic acid, 4-ethoxy, p-ethoxyphenylacetic acid, para-ethoxyphenylacetic acid, acmc-20amjb, 4-ethoxyphenylaceticacid, enamine_004828, p-ethoxyphenyl acetic acid
PubChem CID 78631
IUPAC Name 2-(4-ethoxyphenyl)acetic acid
SMILES CCOC1=CC=C(C=C1)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 86°C to 90°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5OC6H4CH2CO2H
Quantity 10 g
Beilstein 10, 190
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxyphenylacetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethoxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.