missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl trans-4-(4-n-pentylcyclohexyl)benzoate, 99%, Thermo Scientific™

Product Code. 15482726
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15482726 5 g 5g
15472726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15482726 Supplier Thermo Scientific Alfa Aesar Supplier No. H56493.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95973-51-6
Molecular Formula C26H34O3
Molecular Weight (g/mol) 394.555
MDL Number MFCD09952999
InChI Key BOJBRLBJBXWHNZ-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl 4-trans-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 2-trans-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 4-1s,4r-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl 4-4-pentylcyclohexyl benzoate, 4-ethoxyphenyl-4'-trans-pentylcyclohexyl benzoate, 4-ethoxyphenyl trans-4-4-n-pentylcyclohexyl benzoate
PubChem CID 23334669
IUPAC Name (4-ethoxyphenyl) 4-(4-pentylcyclohexyl)benzoate
SMILES CCCCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)OC3=CC=C(C=C3)OCC

Specifications

Quantity 5 g
Formula Weight 394.55
Percent Purity 99%
Chemical Name or Material 4-Ethoxyphenyl trans-4-(4-n-pentylcyclohexyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.