missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isothiocyanate 97%, Maybridge™

Brand:  Maybridge SB01663EA

Code : 31

Additional Details : CAS Number : 3460-49-9 Weight : 0.06000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10596413

  • £20.09 / 10g

Estimated Shipment
07-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

3460-49-9
179.237
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
1-ethoxy-4-isothiocyanatobenzene
C9H9NOS
REEKOMRZYJXXNR-UHFFFAOYSA-N
77006
CCOC1=CC=C(C=C1)N=C=S

Specifications

4-Ethoxyphenyl isothiocyanate
10g
97%
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
CCOC1=CC=C(C=C1)N=C=S
179.237
179.24
Amber glass bottle
3460-49-9
C9H9NOS
REEKOMRZYJXXNR-UHFFFAOYSA-N
1-ethoxy-4-isothiocyanatobenzene
77006
97%
Videos
SDS Documents