missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxyphenyl isothiocyanate 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 25 g |
Formula Weight | 179.24 |
Percent Purity | 97% |
Chemical Name or Material | 4-Ethoxyphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'