Learn More
4-Ethoxyphenyl isothiocyanate 97%, Maybridge™
Brand: Maybridge SB01663DE
Code : 31
Additional Details : CAS Number : 3460-49-9 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
3460-49-9 | |
179.237 | |
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate | |
1-ethoxy-4-isothiocyanatobenzene |
C9H9NOS | |
REEKOMRZYJXXNR-UHFFFAOYSA-N | |
77006 | |
CCOC1=CC=C(C=C1)N=C=S |
Specifications
4-Ethoxyphenyl isothiocyanate | |
5g | |
97% | |
4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate | |
CCOC1=CC=C(C=C1)N=C=S | |
179.237 | |
179.24 |
Amber glass bottle | |
3460-49-9 | |
C9H9NOS | |
REEKOMRZYJXXNR-UHFFFAOYSA-N | |
1-ethoxy-4-isothiocyanatobenzene | |
77006 | |
97% |