missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isocyanate, 97%

Product Code. 11392457
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392457 25 g 25g
11382457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392457 Supplier Thermo Scientific Alfa Aesar Supplier No. L11509.14
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32459-62-4 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32459-62-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00013877
InChI Key FMYVTFRADSNGDN-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isocyanate, benzene, 1-ethoxy-4-isocyanato, 4-ethoxyphenylisocyanate, 4-isocyanatophenetole, isocyanic acid 4-ethoxyphenyl ester, p-ethoxy phenyl isocyanate, 4-ethoxybenzenisocyanate, pubchem3178, p-athoxyphenylisocyanat, acmc-1afep
PubChem CID 122887
IUPAC Name 1-ethoxy-4-isocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=O

Specifications

Density 1.11
Boiling Point 235°C
Flash Point 105°C (221°F)
Refractive Index 1.536
Quantity 25 g
UN Number UN2206
Beilstein 742647
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 4-Ethoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.