missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenol, 99%

Product Code. 11438987
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438987 50 g 50g
11428987 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438987 Supplier Thermo Scientific Alfa Aesar Supplier No. B20584.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 622-62-8 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-62-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002334
InChI Key LKVFCSWBKOVHAH-UHFFFAOYSA-N
Synonym phenol, 4-ethoxy, p-ethoxyphenol, hydroquinone monoethyl ether, 4-ethyloxyphenol, p-hydroxyphenetole, phenol, p-ethoxy, 4-ethoxy-phenol, 1-ethoxy-4-hydroxybenzene, para-ethoxyphenol, ethoxyphenol, p
PubChem CID 12150
IUPAC Name 4-ethoxyphenol
SMILES CCOC1=CC=C(C=C1)O

Specifications

Melting Point 64°C to 67°C
Boiling Point 121°C (9 mmHg)
Odor Phenol-like
Quantity 50 g
Beilstein 1907114
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 4-Ethoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.