missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenol, 99%

Product Code. 10299033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299033 100 g Glass bottle 100g
10670925 5 g Glass bottle 5g
10055720 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299033 Supplier Thermo Scientific Acros Supplier No. 409671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-62-8 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-62-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002334
InChI Key LKVFCSWBKOVHAH-UHFFFAOYSA-N
Synonym phenol, 4-ethoxy, p-ethoxyphenol, hydroquinone monoethyl ether, 4-ethyloxyphenol, p-hydroxyphenetole, phenol, p-ethoxy, 4-ethoxy-phenol, 1-ethoxy-4-hydroxybenzene, para-ethoxyphenol, ethoxyphenol, p
PubChem CID 12150
IUPAC Name 4-ethoxyphenol
SMILES CCOC1=CC=C(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 67°C
Color Beige
Boiling Point 131°C (9.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,ethyl ether,soluble in chloroform
Formula Weight 138.17
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Ethoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethoxyphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.