missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycarbonyl-3-ethoxyphenylacetic Acid, TRC

Product Code. 30384705
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384705 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384705 Supplier Toronto Research Chemicals Supplier No. TRCE890470500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic Acid
CAS 99469-99-5
Molecular Formula C13 H16 O5
Quantity 500 mg
Synonym [3-Ethoxy-4-(ethoxycarbonyl)phenyl]acetic Acid, Repaglinide Imp. B (EP)
InChI Formula InChI=1S/C13H16O5/c1-3-17-11-7-9(8-12(14)15)5-6-10(11)13(16)18-4-2/h5-7H,3-4,8H2,1-2H3,(H,14,15)
SMILES CCOC(=O)c1ccc(CC(=O)O)cc1OCC
IUPAC Name 2-(3-ethoxy-4-ethoxycarbonylphenyl)acetic acid
Molecular Weight (g/mol) 252.26
Formula Weight 252.0998
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.