missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 15451916
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451916 250 mg 250mg
15441916 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451916 Supplier Thermo Scientific Alfa Aesar Supplier No. H55087.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851334-92-4
Molecular Formula C15H20BFO4
Molecular Weight (g/mol) 294.129
MDL Number MFCD18383988
InChI Key ZXSGWPHDXQZIAF-UHFFFAOYSA-N
Synonym ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-ethoxycarbonyl-2-fluorophenylboronic acid, pinacol ester, 4-ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester, ethyl 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzoate, ethyl 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, ethyl3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 23134230
IUPAC Name ethyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)OCC)F

Specifications

Quantity 250 mg
Beilstein 14562502
Formula Weight 294.13
Percent Purity 97%
Chemical Name or Material 4-Ethoxycarbonyl-2-fluorobenzeneboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.