missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzyl alcohol, 98%

Product Code. 11454103
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454103 10 g 10g
11464103 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454103 Supplier Thermo Scientific Alfa Aesar Supplier No. A10331.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6214-44-4 | C9H12O2 | 152.19 g/mol

It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6214-44-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004655
InChI Key UKFLLQIRBABMKF-UHFFFAOYSA-N
Synonym 4-ethoxybenzyl alcohol, 4-ethoxyphenyl methanol, p-ethoxybenzyl alcohol, 4-ethoxybenzylalcohol, benzenemethanol, 4-ethoxy, 4-ethoxy-benzyl alcohol, benzenemethanol,4-ethoxy, 4-ethoxyphenyl methanol #, 4-ethoxyphenyl methan-1-ol, 4-ethyloxy phenyl methanol
PubChem CID 80345
IUPAC Name (4-ethoxyphenyl)methanol
SMILES CCOC1=CC=C(CO)C=C1

Specifications

Melting Point 32°C to 34°C
Boiling Point 272°C to 274°C
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 10 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.