missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzoic acid, 98+%

Product Code. 11465053
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465053 25 g 25g
11475053 100 g 100g
11485053 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465053 Supplier Thermo Scientific Alfa Aesar Supplier No. A12309.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 619-86-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-86-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002545
InChI Key SHSGDXCJYVZFTP-UHFFFAOYSA-N
Synonym p-ethoxybenzoic acid, benzoic acid, 4-ethoxy, benzoic acid, p-ethoxy, 4-ethoxy-benzoic acid, unii-tzr23xeq7w, tzr23xeq7w, 4-ethoxybenzoicacid, 4ethoxybenzoic acid, pubchem2686, para-ethoxybenzoic acid
PubChem CID 12093
IUPAC Name 4-ethoxybenzoic acid
SMILES CCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 197°C to 199°C
Quantity 25 g
Beilstein 2207349
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxybenzoic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.