missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzaldehyde, 97+%

Product Code. 11494024
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494024 500 g 500g
11474024 25 g 25g
11484024 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494024 Supplier Thermo Scientific Alfa Aesar Supplier No. A11987.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-82-0 | C9H10O2 | 150.18 g/mol

4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethoxybenzaldehyde can be used as an anti-inflammatory and prostaglandin E2 suppressor. benzaldehyde-derived anti-inflammatory agents may be useful in reducing facial erythema.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents. Air sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-82-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00003388
InChI Key JRHHJNMASOIRDS-UHFFFAOYSA-N
Synonym benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
PubChem CID 24834
IUPAC Name 4-ethoxybenzaldehyde
SMILES CCOC1=CC=C(C=O)C=C1

Specifications

Melting Point 13°C to 14°C
Density 1.08
Boiling Point 253°C to 255°C
Flash Point <110°C (230°F)
Refractive Index 1.559
Quantity 500 g
Beilstein 386863
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity ≥97%
Chemical Name or Material 4-Ethoxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.