missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethoxyacetophenone, 99%

Product Code. p-3736409 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10058931 5 g 5g
10444615 25 g 25g
10187130 100 g 100g
11944881 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10058931 Supplier Thermo Scientific Acros Supplier No. 409590050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1676-63-7 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00009095
InChI Key YJFNFQHMQJCPRG-UHFFFAOYSA-N
Synonym 4'-ethoxyacetophenone, 1-4-ethoxyphenyl ethanone, p-ethoxyacetophenone, 4-ethoxyacetophenone, 1-4-ethoxyphenyl ethan-1-one, ethanone, 1-4-ethoxyphenyl, 4'-ethoxy acetophenone, acetophenone, p-ethoxy, acetophenone, 4'-ethoxy, 4-ethoxyphenylethanone
PubChem CID 72872
IUPAC Name 1-(4-ethoxyphenyl)ethanone
SMILES CCOC1=CC=C(C=C1)C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 35.0°C to 38.0°C
Boiling Point 268.0°C to 269.0°C (758.0 mmHg)
Flash Point 134°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 4'-Ethoxyacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Ethoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.