missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-N,N,N-trimethyl-2,4-dioxo-1-butanaminium Chloride, TRC

Product Code. 30376782
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30376782 500 mg
30388260 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30376782 Supplier Toronto Research Chemicals Supplier No. TRCE892985500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10485-23-1
Molecular Formula C9H18NO3 • Cl
Molecular Weight (g/mol) 223.7
Synonym (3-Carboxyacetonyl)trimethylammonium Chloride Ethyl Ester, (3-Carboxy-2-hydroxyallyl)trimethyl-ammonium Chloride Ethyl Ester
IUPAC Name 4-ethoxy-N,N,N-trimethyl-2,4-dioxobutan-1-aminium chloride
SMILES [Cl-].O=C(OCC)CC(=O)C[N+](C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H18NO3.ClH/c1-5-13-9(12)6-8(11)7-10(2,3)4;/h5-7H2,1-4H3;1H/q+1;/p-1
Formula Weight 224.1
Chemical Name or Material 4-Ethoxy-N,N,N-trimethyl-2,4-dioxo-1-butanaminium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.