missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-methoxybenzaldehyde, 95%, Thermo Scientific™

Product Code. 10634962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10634962 25 g Plastic bottle 25g
10725463 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10634962 Supplier Thermo Scientific Acros Supplier No. 117910250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 120-25-2
MDL Number MFCD00016614
InChI Key BERFDQAMXIBOHM-UHFFFAOYSA-N
Synonym 4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648
PubChem CID 67116
IUPAC Name 4-ethoxy-3-methoxybenzaldehyde
SMILES CCOC1=C(C=C(C=C1)C=O)OC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 60.0°C to 65.0°C
Color Yellow
Boiling Point 168.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Molecular Formula C10H12O3
Linear Formula C2H5OC6H3(OCH3)CHO
Quantity 25 g
Beilstein 08,256
Solubility Information Solubility in water: slightly soluble
Molecular Weight (g/mol) 180.2
Formula Weight 180.2
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.