missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2-fluorobenzamide, 97%, Thermo Scientific™

Catalog Number 15416777
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1206593-31-8
C9H10FNO2
183.182
MFCD13196592
ZSPVCLBFKSZFIP-UHFFFAOYSA-N
57361365
4-ethoxy-2-fluorobenzamide
CCOC1=CC(=C(C=C1)C(=O)N)F
This item is not returnable. View return policy
1206593-31-8
C9H10FNO2
183.182
MFCD13196592
ZSPVCLBFKSZFIP-UHFFFAOYSA-N
57361365
4-ethoxy-2-fluorobenzamide
CCOC1=CC(=C(C=C1)C(=O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 183.18
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2-fluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'