missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 15497027
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15497027 1 g 1g
15407037 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15497027

Brand: Thermo Scientific Alfa Aesar H34113.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373920-79-6
Molecular Formula C9H7ClF2O2
Molecular Weight (g/mol) 220.6
MDL Number MFCD22201053
InChI Key DFCDPQTYYGJYOD-UHFFFAOYSA-N
PubChem CID 86277653
IUPAC Name 4-ethoxy-2,6-difluorobenzoyl chloride
SMILES CCOC1=CC(=C(C(=C1)F)C(=O)Cl)F

Specifications

Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.