missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluoroaniline, 97%, Thermo Scientific™

Product Code. 15444347
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1017779-79-1
Molecular Formula C8H9F2NO
Refractive Index 1.5084
MDL Number MFCD09832394
Quantity 250 mg
UN Number UN2810
Synonym 2,6-difluoro-p-phenetidine, 4-ethoxy-2,6-difluorophenylamine, 4-ethoxy-2,6-difluoroaniline, jrd
InChI Key WEVYXUJTENEFCM-UHFFFAOYSA-N
SMILES CCOC1=CC(F)=C(N)C(F)=C1
IUPAC Name 4-ethoxy-2,6-difluoroaniline
Molecular Weight (g/mol) 173.16
PubChem CID 20112218
Formula Weight 173.16
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,6-difluoroaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'