missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,3-difluorobenzoic acid, 98%, Thermo Scientific™

Catalog Number p-7028685
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
124728-45-6
C9H8F2O3
202.157
MFCD02258849
KMFHBAZUQFJVKK-UHFFFAOYSA-N
2769400
4-ethoxy-2,3-difluorobenzoic acid
CCOC1=C(C(=C(C=C1)C(=O)O)F)F
This item is not returnable. View return policy
124728-45-6
C9H8F2O3
202.157
MFCD02258849
KMFHBAZUQFJVKK-UHFFFAOYSA-N
2769400
4-ethoxy-2,3-difluorobenzoic acid
CCOC1=C(C(=C(C=C1)C(=O)O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 200°C to 203°C
Quantity 250 mg
Synonym 2,3-difluoro-4-ethoxybenzoic acid, benzoic acid, 4-ethoxy-2,3-difluoro, acmc-20aari, 4-carboxy-2,3-difluorophenetole, benzoic acid,4-ethoxy-2,3-difluoro, benzoic acid, 4-ethoxy-2,3-difluoro-9ci
Formula Weight 202.15
Percent Purity 98%
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'