missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- 4-Epianhydrotetracycline hydrochloride is a secondary degradation product formed by epimerization of tetracycline and dehydration at the C6 position to aromatize the B ring
- It has 250-fold higher toxicity than either tetracycline or doxycycline depending on the system under study
Applications
- 4-Epianhydrotetracycline hydrochloride is less toxic to E. coli, and thus suitable for use with vectors or systems designed upon elements of the (Tn10) tet operon of E. coli
- This compound is also used in veterinary medicine research and as a secondary standard for application in quality control in pharmaceutical laboratories
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 220.0°C |
| Color | Orange |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 80% min. (as is,calculated) |
| Packaging | Glass bottle |
| Quantity | 500 mg |
| Specific Rotation | − 279.00 |
| Solubility Information | Solubility in water: 0.9g/L. Other solubilities: 25g/L methanol |
| Show More |
Safety and Handling
Product Identifier
- 4-Epianhydrotetracycline hydrochloride
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 2
- H319-Causes serious eye irritation.
- H361d-Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction