missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-Epianhydrotetracycline hydrochloride, can be used as a secondary standard

Product Code. 10073170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10073170 100 mg Glass bottle 100mg
10751891 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073170 Supplier Thermo Scientific Chemicals Supplier No. 233111000
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4-Epianhydrotetracycline hydrochloride, CAS # 4465-65-0, is an antibiotic derivative of tetracycline. It is an important standard for monitoring tetracycline stability and toxicity in laboratory studies.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • 4-Epianhydrotetracycline hydrochloride is a secondary degradation product formed by epimerization of tetracycline and dehydration at the C6 position to aromatize the B ring
  • It has 250-fold higher toxicity than either tetracycline or doxycycline depending on the system under study

Applications

  • 4-Epianhydrotetracycline hydrochloride is less toxic to E. coli, and thus suitable for use with vectors or systems designed upon elements of the (Tn10) tet operon of E. coli
  • This compound is also used in veterinary medicine research and as a secondary standard for application in quality control in pharmaceutical laboratories
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4465-65-0
Molecular Formula C22H22N2O7·HCl
Molecular Weight (g/mol) 462.89
MDL Number MFCD00151455
InChI Key SPFAOPCHYIJPHJ-MOMXNFOMSA-N
Synonym 4-epianhydrotetracycline hydrochloride, unii-06y7hm8dba, 06y7hm8dba, 4-epi-anhydrotetracycline hydrochloride, 4-epi-ahydrochloride, 4-epianhydrotetracycline hcl, epianhydrotetracycline hydrochloride, 4-epianhydrotetracycline hydrochloride, vetranal tm , analytical standard, 4-epianhydrotetracycline hydrochloride, european pharmacopoeia ep reference standard, 4-epianhydrotetracycline hydrochloride, united states pharmacopeia usp reference standard
PubChem CID 54682545
IUPAC Name (4R,4aS,12aR)-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 100 mg
Specific Rotation − 279.00
Solubility Information Solubility in water: 0.9g/L. Other solubilities: 25g/L methanol
Formula Weight 462.89
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Powder
Chemical Name or Material 4-Epianhydrotetracycline hydrochloride, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Epianhydrotetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.