missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecanol, 99%, Thermo Scientific Chemicals

Product Code. 11367895
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367895 5 g 5g
11377895 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367895 Supplier Thermo Scientific Alfa Aesar Supplier No. L02921.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10203-32-4
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00014413
InChI Key ZGSIAHIBHSEKPB-UHFFFAOYNA-N
Synonym 4-dodecanol, 4-hydroxydodecane, acmc-1c0r3
PubChem CID 66291
IUPAC Name dodecan-4-ol
SMILES CCCCCCCCC(O)CCC

Specifications

Melting Point 11°C to 12°C
Density 0.829
Boiling Point 130°C to 132°C (19 mmHg)
Refractive Index 1.44
Quantity 5 g
Beilstein 1738597
Formula Weight 186.34
Percent Purity 99%
Chemical Name or Material 4-Dodecanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.