missing translation for 'onlineSavingsMsg'
Learn More

4-[(Dipropylamino)sulfonyl]benzoic acid, 97%, Maybridge™

Product Code. 10503145
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13H19NO4S
Molecular Weight (g/mol) 285.36
MDL Number MFCD00038402
InChI Key DBABZHXKTCFAPX-UHFFFAOYSA-N
Synonym probenecid, 4-dipropylsulfamoyl benzoic acid, probenecid acid, probecid, benecid, benemid, benuryl, probalan, probenemid, probexin
PubChem CID 4911
ChEBI CHEBI:8426
IUPAC Name 4-(dipropylsulfamoyl)benzoic acid
SMILES CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula [CH3(CH2)2]2NSO2C6H4CO2H
Quantity 25 g
Formula Weight 285.36
Percent Purity 97%
Chemical Name or Material 4-[(Dipropylamino)sulfonyl]benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'