missing translation for 'onlineSavingsMsg'
Learn More

4-Diphenylmethoxypiperidine hydrochloride, TRC

Product Code. 30381633
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381633 25 mg
30374955 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381633 Supplier Toronto Research Chemicals Supplier No. TRCD46267025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65214-86-0
Molecular Formula C18 H21 N O . Cl H
Molecular Weight (g/mol) 303.83
Synonym N-Desmethyldiphenylpyraline Hydrochloride, 4-(Diphenylmethoxy)piperidine Hydrochloride, Ebastine Imp. C (EP) as Hydrochloride
IUPAC Name 4-benzhydryloxypiperidine;hydrochloride
SMILES Cl.C1CC(CCN1)OC(c2ccccc2)c3ccccc3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H21NO.ClH/c1-3-7-15(8-4-1)18(16-9-5-2-6-10-16)20-17-11-13-19-14-12-17;/h1-10,17-19H,11-14H2;1H
Formula Weight 303.139
Chemical Name or Material 4-(Diphenylmethoxy)piperidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.