missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Diphenylmethoxy)piperidine Hydrochloride, Mikromol™
Description
4-(Diphenylmethoxy)piperidine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 4-(Diphenylmethoxy)piperidine Hydrochloride |
| CAS | 65214-86-0 |
| Analyte or Component Names | 4-(Diphenylmethoxy)piperidine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H21 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ebastine Imp. C (EP), N-Desmethyldiphenylpyraline Hydrochloride, 4-(Diphenylmethoxy)piperidine Hydrochloride, Ebastine Imp. C (EP) as Hydrochloride, Ebastine Related Compound C as Hydrochloride, Ebastine Impurity C as Hydrochloride, 4-(benzhydryloxy)piperidine hydrochloride, Piperidine, 4-(diphenylmethoxy)-, hydrochloride (1:1) (ACI), Piperidine, 4-(diphenylmethoxy)-, hydrochloride (9CI) |
| InChI Formula | InChI=1S/C18H21NO.ClH/c1-3-7-15(8-4-1)18(16-9-5-2-6-10-16)20-17-11-13-19-14-12-17;/h1-10,17-19H,11-14H2;1H |
| SMILES | Cl.C1CC(CCN1)OC(c2ccccc2)c3ccccc3 |
| IUPAC Name | 4-benzhydryloxypiperidine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?