missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzeneboronic acid pinacol ester, 95%

Product Code. 15439455
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439455 1 g 1g
15449455 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439455 Supplier Thermo Scientific Alfa Aesar Supplier No. H51946.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 267221-88-5 | C24H26BNO2 | 371.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 267221-88-5
Molecular Formula C24H26BNO2
Molecular Weight (g/mol) 371.29
MDL Number MFCD13195770
InChI Key VKSWIFGDKIEVFZ-UHFFFAOYSA-N
Synonym n,n-diphenyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 4-diphenylamino phenylboronic acid pinacol ester, diphenyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl amine, amtb248, 4-diphenylamino phenylboronic acid, pinacol ester, 4-diphenylamino benzeneboronic acid pinacol ester, n,n-diphenylaniline-4-boronic acid, pinacol ester, 2-4-diphenylaminophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-4-diphenylaminophenyl-1,3,2-dioxaborolane
PubChem CID 11639307
IUPAC Name N,N-diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Beilstein 10571706
Formula Weight 371.29
Percent Purity 95%
Chemical Name or Material 4-(Diphenylamino)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.