missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylsulfamoyl)benzene-1-sulfonyl Chloride, TRC

Product Code. 30385915
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385915 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385915 Supplier Toronto Research Chemicals Supplier No. TRCD48454510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Dimethylsulfamoyl)benzene-1-sulfonyl Chloride
CAS 677782-39-7
Molecular Formula C8 H10 Cl N O4 S2
Quantity 10 mg
Synonym 4-[(Dimethylamino)sulfonyl]benzenesulfonyl chloride, 4-(Dimethylsulfamoyl)benzenesulfonyl chloride
InChI Formula InChI=1S/C8H10ClNO4S2/c1-10(2)16(13,14)8-5-3-7(4-6-8)15(9,11)12/h3-6H,1-2H3
SMILES CN(C)S(=O)(=O)c1ccc(cc1)S(=O)(=O)Cl
IUPAC Name 4-(dimethylsulfamoyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 283.752
Formula Weight 282.974
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.