missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15430396
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
874289-13-1
C9H11BFNO3
210.999
MFCD08436060
KEZPQOGKDGNRHH-UHFFFAOYSA-N
4-dimethylcarbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorobenzeneboronic acid, acmc-209qm6, 4-dimethylcarbamoyl-3-fluorophenyl boronicacid, 4-dimethyl carbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluoranyl-phenyl boronic acid, 4-dimethylamino oxo methyl-3-fluorophenyl boronic acid, boronic acid,b-4-dimethylamino carbonyl-3-fluorophenyl
44717474
[4-(dimethylcarbamoyl)-3-fluorophenyl]boronic acid
B(C1=CC(=C(C=C1)C(=O)N(C)C)F)(O)O
This item is not returnable. View return policy
874289-13-1
C9H11BFNO3
210.999
MFCD08436060
KEZPQOGKDGNRHH-UHFFFAOYSA-N
4-dimethylcarbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluorophenyl boronic acid, 4-dimethylcarbamoyl-3-fluorobenzeneboronic acid, acmc-209qm6, 4-dimethylcarbamoyl-3-fluorophenyl boronicacid, 4-dimethyl carbamoyl-3-fluorophenylboronic acid, 4-dimethylcarbamoyl-3-fluoranyl-phenyl boronic acid, 4-dimethylamino oxo methyl-3-fluorophenyl boronic acid, boronic acid,b-4-dimethylamino carbonyl-3-fluorophenyl
44717474
[4-(dimethylcarbamoyl)-3-fluorophenyl]boronic acid
B(C1=CC(=C(C=C1)C(=O)N(C)C)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 149°C
Quantity 250 mg
Formula Weight 211
Percent Purity 98%
Chemical Name or Material 4-Dimethylcarbamoyl-3-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'