missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, TRC

Product Code. 30010654
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010654 25 g 25g
30011203 250 g 250g
30004988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010654 Supplier Toronto Research Chemicals Supplier No. TRCD47902525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-58-3
Molecular Formula C7 H10 N2
Molecular Weight (g/mol) 122.17
Synonym Pyridine, 4-(dimethylamino)- (6CI,7CI,8CI), N,N-Dimethyl-4-pyridinamine, 4-(Dimethylamino)pyridine, 4-(N,N-Dimethylamino)pyridine, 4-DMAP, DMAP, DMAP (catalyst), N,N-Dimethyl-4-aminopyridine, N,N-Dimethyl-4-pyridylamine, p-Dimethylaminopyridine, γ-(Dimethylamino)pyridine
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)c1ccncc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H10N2/c1-9(2)7-3-5-8-6-4-7/h3-6H,1-2H3
Formula Weight 122.0844
Percent Purity >95%
Chemical Name or Material 4-Dimethylaminopyridine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.