missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylaminobenzylidene)rhodanine, TRC

Product Code. 30383358
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383358 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383358 Supplier Toronto Research Chemicals Supplier No. TRCD8265932.5G
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Dimethylaminobenzylidene)rhodanine
CAS 536-17-4
Molecular Formula C12 H12 N2 O S2
Quantity 2.5 g
Synonym 5-(4-Dimethylaminobenzylidene)rhodanine
InChI Formula InChI=1S/C12H12N2OS2/c1-14(2)9-5-3-8(4-6-9)7-10-11(15)13-12(16)17-10/h3-7H,1-2H3,(H,13,15,16)
SMILES CN(C)c1ccc(C=C2SC(=S)NC2=O)cc1
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol) 264.366
Formula Weight 264.039
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.