missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, ACS reagent

Product Code. 10700302 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10700302 500 g Glass bottle 500g
10138230 25 g Glass bottle 25g
10697031 100 g Glass bottle 100g
10627954 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10700302 Supplier Thermo Scientific Acros Supplier No. 423635000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-10-7 | C9H11NO | 149.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 70°C to 75°C
Color White to Off-white
Density 5.15
Boiling Point 176°C to 177°C (17.0 mmHg)
Flash Point 164°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 500 g
Beilstein 14, 31
Fieser 01,273; 02,146
Merck Index 15, 3252
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in alcohol,ether and chloroform
Formula Weight 149.19
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.