missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)tetrahydro-2H-pyran-4-carbonitrile, 97%, Thermo Scientific™

Catalog Number 10326814
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
176445-77-5
C8H14N2O
154.21
MFCD09065026
FDGOISRHLNMQRE-UHFFFAOYSA-N
4-dimethylamino tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino oxane-4-carbonitrile, 4-cyano-4-dimethylamino-tetrahydro-4h-pyrane, 2h-pyran-4-carbonitrile,4-dimethylamino tetrahydro, 4-dimethylamino-tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino-2h-3,4,5,6-tetrahydropyran-4-carbonitrile
10511078
4-(dimethylamino)oxane-4-carbonitrile
CN(C)C1(CCOCC1)C#N
This item is not returnable. View return policy
176445-77-5
C8H14N2O
154.21
MFCD09065026
FDGOISRHLNMQRE-UHFFFAOYSA-N
4-dimethylamino tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino oxane-4-carbonitrile, 4-cyano-4-dimethylamino-tetrahydro-4h-pyrane, 2h-pyran-4-carbonitrile,4-dimethylamino tetrahydro, 4-dimethylamino-tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino-2h-3,4,5,6-tetrahydropyran-4-carbonitrile
10511078
4-(dimethylamino)oxane-4-carbonitrile
CN(C)C1(CCOCC1)C#N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)tetrahydro-2H-pyran-4-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'