missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)pyridine, 99%

Product Code. p-7020905
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11499923

Brand: Thermo Scientific Alfa Aesar A13016.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-58-3 | C7H10N2 | 122.171 g/mol

4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Dimethylamino)pyridine is a useful nucleophilic catalyst for various reactions viz. esterifications with anhydrides, the Baylis-Hillman reaction, hydrosilylations, tritylation and Steglich rearrangement. It is also an effective catalyst for transesterification of beta-keto esters and silylation of alcohols.

Solubility
Soluble in methanol, benzene, ethyl acetate, chloroform, methylene chloride, acetone, and acetic acid. Slightly soluble in ether, diisopropyl ether, cyclohexane, hexane, and water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-58-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1

Specifications

Melting Point 110°C to 113°C
Boiling Point 162°C (50 mmHg)
Flash Point 124°C (255°F)
Assay Percent Range 99%
Quantity 5 g
UN Number UN2928
Beilstein 110354
Solubility Information Soluble in methanol,benzene,ethyl acetate,chloroform,methylene chloride,acetone,and acetic acid. Slightly soluble in ether,diisopropyl ether,cyclohexane,hexane,and water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 4-(Dimethylamino)pyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-(Dimethylamino)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-(Dimethylamino)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.