missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)piperidine, 97%, Thermo Scientific Chemicals
173.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
Notes
Incompatible with air, water, oxidizing agents and heat.
4-(Dimethylamino)piperidine is having molecular features associated with polyamine modulation of N-methyl-D-aspartate receptors. It is utilized as a mechanism for reducing the effects of alcohol dependence.
Notes
Incompatible with air, water, oxidizing agents and heat.
Specifications
Boiling Point | 187°C |
Refractive Index | 1.476 |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 1190 |
Formula Weight | 128.22 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Dimethylamino)piperidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'