missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzonitrile, 95%

Product Code. 10254140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10254140 5 g Glass bottle 5g
10756141 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10254140 Supplier Thermo Scientific Acros Supplier No. 115780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1197-19-9 | C9H10N2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1197-19-9
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001815
InChI Key JYMNQRQQBJIMCV-UHFFFAOYSA-N
Synonym 4-dimethylamino benzonitrile, p-dimethylaminobenzonitrile, benzonitrile, 4-dimethylamino, n,n-dimethyl-p-cyanoaniline, p-cyano-n,n-dimethylaniline, 4-cyano-n,n-dimethylaniline, n,n-dimethyl-4-cyanoaniline, 4-dimethylamino benzenecarbonitrile, para dimethylamino benzonitrile, 4-cyano-nn-dimethylaniline
PubChem CID 70967
IUPAC Name 4-(dimethylamino)benzonitrile
SMILES CN(C)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 70.0°C to 76.0°C
Color Brown
Boiling Point 318.0°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4CN
Quantity 5 g
Beilstein 14, 428
Formula Weight 146.19
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)benzonitrile, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.